Learn More
Aviva Systems Biology Corp SV2A Antibody - middle region (ARP47093_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47093_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SV2A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SV2A. Alternate names: KIAA0736, SV2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53504