Learn More
Coast SV400 High-Vis Glow-in-the-Dark Safety Vest with Removable LED Strip
Catalog No. p-7217104
Change view
Click to view available options
Size:
Large
Catalog No. Size
19-012-105 Large
1 options
This item is not returnable. View return policy
Catalog No. 19-012-105 Supplier Coast Supplier No. 30390
This item is not returnable. View return policy
ANSI Class 2 Vests are constructed of high-vis yellow mesh fabric, and enhanced by bands of glow-in-the-dark phosphorescent fabric quickly charge under any light source.
  • Two-tone high-vis safety vests are made with ANSI Class 2 high-vis yellow mesh fabric
  • Enhanced by bands of glow-in-the-dark phosphorescent fabric that quickly charge under any light source
  • Provides long-lasting, automatic lighting when transitioning from bright to dark environments
  • A rechargeable, removable LED safety light strip on the upper back further enhances safety with solid and flashing light modes in safety orange
  • Vest features two shoulder-mounted mic tabs, a behind-the-neck retroreflective tab for clip-on safety lights, an oversized inner chest pocket, and an outer chest pocket
  • Zipper front closure for a secure fit
  • Washable
Specifications
Closure Type Zipper
Certifications/Compliance ANSI 107-2015 Class 2 Type R
Color Hi-vis Yellow
Size Large
Material Polyester Mesh
Includes Removable LED Light Strip
No. of Pockets 2
Tape Type Reflective