Learn More
Enzo Life Sciences sVEGFR-1 (human) ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ALX850264KI01
Encompass_Preferred
The sVEGFR-1 (human) ELISA kit is a complete assay for the quantitative determination of sVEGFR-1 levels. This kit is to be used with cell culture supernatants, serum and other body fluids from human origin. Alternative name: Vascular endothelial growth factor receptor 1, Flt-1. Assay Time: 3.5 hours. Applications: ELISA . Species reactivity: Human .
Catalog No. 50-201-0145