Learn More
ATCC SV-HUC-1; Uroepithelium; Human (Homo sapiens)
Catalog No. 502384971
Encompass
SV-HUC-1 is an epithelial cell isolated from the uroepithelium of an 11-year-old, male. This line was established by the transformation of normal ureter tissue with the SV40 virus.
Catalog No. 50-238-4971 Supplier ATCC Supplier No. CRL9520
SV-HUC-1 is an epithelial cell isolated from the uroepithelium of an 11-year-old, male. This line was established by the transformation of normal ureter tissue with the SV40 virus.