Learn More
Medchemexpress LLC HY-115475 5mg Medchemexpress, SW-100 CAS:2126744-35-0 Purity:>98%
SDP
Catalog No. 501932504
Encompass_Preferred
Medchemexpress, HY-115475 5mg SW-100 CAS:2126744-35-0 SW-100, a selective histone deacetylase 6 (HDAC6) inhibitor with an IC 50 of 2.3 nM, shows at least 1000-fold selectivity for HDAC6 relative to all other HDAC isozymes. SW-100 displays a significantly improved ability to cross the blood-brain-barrier [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2504 Supplier Medchemexpress LLC Supplier No. HY1154755MG
Medchemexpress, HY-115475 5mg SW-100 CAS:2126744-35-0 SW-100, a selective histone deacetylase 6 (HDAC6) inhibitor with an IC 50 of 2.3 nM, shows at least 1000-fold selectivity for HDAC6 relative to all other HDAC isozymes. SW-100 displays a significantly improved ability to cross the blood-brain-barrier [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.