Learn More
ATCC SW 1990; Pancreatic Carcinoma; Human (Homo sapiens)
Catalog No. 502381740
Encompass
SW 1990 [SW-1990, SW1990] epithelial cells were isolated from the spleen metastasis of a grade II pancreatic adenocarcinoma derived from the exocrine pancreas of a 56-year-old, White male in 1978. Use the cell line in your cancer research.
Catalog No. 50-238-1740 Supplier ATCC Supplier No. CRL2172
SW 1990 [SW-1990, SW1990] epithelial cells were isolated from the spleen metastasis of a grade II pancreatic adenocarcinoma derived from the exocrine pancreas of a 56-year-old, White male in 1978. Use the cell line in your cancer research.