Learn More
ATCC SW 872; Liposarcoma; Human (Homo sapiens)
Supplier:  ATCC HTB92
Encompass
SW 872 [SW-872, SW872] is a cell line exhibiting fibroblast morphology that was isolated from the connective tissue of a 36-year-old, White male patient with liposarcoma. This product has applications in cancer research.
Catalog No. 50-238-2331