Learn More
ATCC SW1353; Chondrosarcoma; Human (Homo sapiens)
Catalog No. 502382614
Encompass
SW 1353 is a fibroblast-like cell isolated from the bone of a 72-year-old, White female with chondrosarcoma.
Catalog No. 50-238-2614 Supplier ATCC Supplier No. HTB94
SW 1353 is a fibroblast-like cell isolated from the bone of a 72-year-old, White female with chondrosarcoma.