Learn More
Medchemexpress LLC Sw2_110a | 00-00-0 | 99.2% | C42H60N6O7 | 10MG
SDP
Catalog No. 5000260763
Encompass_Preferred
SW2_110A is a selective chromobox 8 chromodomain (CBX8 ChD) inhibitor with a reported Kd of 800 nM and cellular activity (IC50 ≈ 26 μM) in MLL-AF9-driven THP1 leukemia cells. It is supplied as a high-purity research compound (molecular formula C42H60N6O7, molecular weight 760.96 g·mol⁻1).
  • Selective CBX8 chromodomain inhibition demonstrated in biochemical assays.
  • Reported binding affinity Kd of 800 nM.
  • Cellular activity with IC50 ≈ 26 μM in THP1 leukemia cells.
  • High purity (99.2%).
  • Supplied as a small-quantity research sample suitable for in vitro studies.
Catalog No. 50-002-60763 Supplier Medchemexpress LLC Supplier No. HY14171610MG
SW2_110A is a selective chromobox 8 chromodomain (CBX8 ChD) inhibitor with a reported Kd of 800 nM and cellular activity (IC50 ≈ 26 μM) in MLL-AF9-driven THP1 leukemia cells. It is supplied as a high-purity research compound (molecular formula C42H60N6O7, molecular weight 760.96 g·mol⁻1).
  • Selective CBX8 chromodomain inhibition demonstrated in biochemical assays.
  • Reported binding affinity Kd of 800 nM.
  • Cellular activity with IC50 ≈ 26 μM in THP1 leukemia cells.
  • High purity (99.2%).
  • Supplied as a small-quantity research sample suitable for in vitro studies.