Learn More
Cayman Chemical SW203668trIfluoROCTAte sa 10mg
SDP
Catalog No. 502982753
Encompass_Preferred
An irreversible inhibitor of stearoyl-CoA desaturase (IC50 = 54 nM); selectively cytotoxic to cancer cell lines that express CYP4F11 over those that don't (IC50s = 22-116 and >10,000 nM, respectively); reduces tumor growth rate without reducing sebocyte production in the H2122 wild-type and NOD-SCID mouse xenograft models at 20 and 6 mg/kg, respectively
Catalog No. 50-298-2753 Supplier Cayman Chemical Supplier No. 1937910 MG
An irreversible inhibitor of stearoyl-CoA desaturase (IC50 = 54 nM); selectively cytotoxic to cancer cell lines that express CYP4F11 over those that don't (IC50s = 22-116 and >10,000 nM, respectively); reduces tumor growth rate without reducing sebocyte production in the H2122 wild-type and NOD-SCID mouse xenograft models at 20 and 6 mg/kg, respectively