Learn More
TARGETMOL CHEMICALS INC SW209049 5MG
SDP
Catalog No. 502578949
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SW209049 is a stearoyl-CoA 9-desaturase inhibitor. SW209049 exhivits potent activity against H2122 cell with IC50 of 0.13uM. purity: 99%
Catalog No. 50-257-8949 Supplier TARGETMOL CHEMICALS INC Supplier No. T98595MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SW209049 is a stearoyl-CoA 9-desaturase inhibitor. SW209049 exhivits potent activity against H2122 cell with IC50 of 0.13uM. purity: 99%