Learn More
Beckman Coulter Beckman Coulter SW 32 Ti Swinging-Bucket Rotor Package (32,000 rpm, 175,000 xg)
Catalog No. NC1812346
Encompass
Beckman Coulter SW 32 Ti Swinging-Bucket Rotor Package is a titanium rotor assembly designed for high-speed ultracentrifugation applications, providing high g-force capability for preparative separations and flexible tube configurations.
  • Maximum speed of 32,000 rpm.
  • Maximum relative centrifugal force of 175,000 xg.
  • k-factor 204.
  • Accepts 6 x 38.5 mL or 25 x 89 mL tubes.
  • Nominal capacity of 231 mL.
  • Titanium construction for high strength and corrosion resistance.
  • Package includes rotor assembly, buckets, seals, lubricant, overspeed disks, and O-rings.
Catalog No. NC1812346 Supplier Beckman Coulter Supplier No. 369694
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
Beckman Coulter SW 32 Ti Swinging-Bucket Rotor Package is a titanium rotor assembly designed for high-speed ultracentrifugation applications, providing high g-force capability for preparative separations and flexible tube configurations.
  • Maximum speed of 32,000 rpm.
  • Maximum relative centrifugal force of 175,000 xg.
  • k-factor 204.
  • Accepts 6 x 38.5 mL or 25 x 89 mL tubes.
  • Nominal capacity of 231 mL.
  • Titanium construction for high strength and corrosion resistance.
  • Package includes rotor assembly, buckets, seals, lubricant, overspeed disks, and O-rings.