Learn More
Amsbio LLC SW48 FFPE Cell Pellet Slide (5-pack)
Catalog No. 502041698
Encompass
FFPE cell line sections are manufactured using a proprietary technology that maintains both cell morphology and preserves nucleic acids and proteins. Available from over 120 tumor cell lines, these FFPE sections make for affordable and consistent controls without compromising quality or consistency. Using cell line controls overcomes issues experienced with traditional cell or tissue-based control specimens such as intra specimen variation, lot-to-lot variations and inconsistent biomolecule quality. Also available in scroll & block formats - please enquire.
Catalog No. 50-204-1698 Supplier Amsbio LLC Supplier No. 305010205PK
FFPE cell line sections are manufactured using a proprietary technology that maintains both cell morphology and preserves nucleic acids and proteins. Available from over 120 tumor cell lines, these FFPE sections make for affordable and consistent controls without compromising quality or consistency. Using cell line controls overcomes issues experienced with traditional cell or tissue-based control specimens such as intra specimen variation, lot-to-lot variations and inconsistent biomolecule quality. Also available in scroll & block formats - please enquire.