Learn More
ATCC SW48 Quantitative; purified DNA, 3 ug; Human
Catalog No. 502383646
Encompass
Purified genomic DNA isolated from the SW-48 human colon adenocarcinoma cell line (ATCC CCL-231). This product was quantified for oncology biomarkers EGFR p.G719S via NGS and ddPCR.
Catalog No. 50-238-3646 Supplier ATCC Supplier No. CCL231DQ
Purified genomic DNA isolated from the SW-48 human colon adenocarcinoma cell line (ATCC CCL-231). This product was quantified for oncology biomarkers EGFR p.G719S via NGS and ddPCR.