Learn More
ATCC SW620 Quantitative; purified DNA, 3 ug; Human
Supplier:  ATCC CCL227DQ
Encompass
Purified genomic DNA isolated from the SW-620 human colon adenocarcinoma cell line (ATCC CCL-227). This product was quantified for oncology biomarkers KRAS p.G12V and TP53 p.R273H via NGS and ddPCR.
Catalog No. 50-238-5020