Learn More
ATCC SW948; Colon Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502381983
Encompass
SW948 is a cell line exhibiting epithelial morphology from the colon of a White, 81-year-old, female patient with adenocarcinoma colorectal Dukes' type C, grade III. This cell line was deposited by A Leibovitz.
Catalog No. 50-238-1983 Supplier ATCC Supplier No. CCL237
SW948 is a cell line exhibiting epithelial morphology from the colon of a White, 81-year-old, female patient with adenocarcinoma colorectal Dukes' type C, grade III. This cell line was deposited by A Leibovitz.