Learn More
Zymo Research Corporation DNA/RNA Shield SafeCollectTM Swab Collection Kit, 2ml
SDP
Catalog No. NC2021762
Encompass_Preferred
User-friendly collection for stabilizing the nucleic acid content of samples collected with a swab. DNA/RNA Shield completely inactivates harmful pathogens in the sample, and stabilizes the RNA in a sample for up to 30 days at room temperature. The SafeCollect design eliminates the risk of spilling the stabilization reagent during use, and is ideal for applications that require at-home self collection.
Catalog No. NC2021762 Supplier Zymo Research Corporation Supplier No. R1161
Add to Cart
Edge
Add to Cart
User-friendly collection for stabilizing the nucleic acid content of samples collected with a swab. DNA/RNA Shield completely inactivates harmful pathogens in the sample, and stabilizes the RNA in a sample for up to 30 days at room temperature. The SafeCollect design eliminates the risk of spilling the stabilization reagent during use, and is ideal for applications that require at-home self collection.