Learn More
Med Vet International Specimen Collection and Transport System ESwab™ Sterile, 220245, Each
Catalog No. 503044208
Encompass_Preferred
Specimen collection and transport system has a flocked swab tip and liquid amies medium additive: It's for aerobic, anaerobic and fastidious bacteria collection Sample rapidly elutes from the flocked swab when placed in the transport medium testing Provides 1 mL of sample suspension for gram stain and multiple culture analyses Suitable platform for automated liquid specimen handling systems
Catalog No. 50-304-4208 Supplier Med Vet International Supplier No. 661468EA
Specimen collection and transport system has a flocked swab tip and liquid amies medium additive: It's for aerobic, anaerobic and fastidious bacteria collection Sample rapidly elutes from the flocked swab when placed in the transport medium testing Provides 1 mL of sample suspension for gram stain and multiple culture analyses Suitable platform for automated liquid specimen handling systems