Learn More
G-Biosciences Swab Collection Kit
Description
Swab collection kit is suitable for sample collection and sample stability during storage and transport at ambient temperatures. Each sample collection tube is pre-filled with 1 mL of conserve buffer that preserves nucleic acids in samples. Swab collection kits also include swabs and biohazard bags.
Specifications
Specifications
| Product Type | Specimen Collection Kit |
| Closure Included | Yes |
| Shape | Conical, Cylindrical |
| Includes | Swab, Collection Tube, 1 mL Conserve Buffer |
| Quantity | 10/Pk. |