Learn More
Air Science Usa Mini Swab Drying Cabinet, Swab Dryer, 115V 60Hz
SDP
Catalog No. NC1360771
Encompass_Preferred
Safe Swab Swab Drying Cabinet, Mini, Benchtop with HEPA main filter formulated to contain any particulates or biological material that might be present on swabs, Safeswab is used to safely dry evidence swabs while protecting them from cross-contamination. Includes one 15 capacity swab drying rack. (fdc-001M)
Catalog No. NC1360771 Supplier Air Science Usa Supplier No. FDC001MA
Safe Swab Swab Drying Cabinet, Mini, Benchtop with HEPA main filter formulated to contain any particulates or biological material that might be present on swabs, Safeswab is used to safely dry evidence swabs while protecting them from cross-contamination. Includes one 15 capacity swab drying rack. (fdc-001M)