Learn More
Med Vet International Impregnated Swabstick 10% Strength Povidone Iodine Individual Packet NonSterile, 1201, 50/Box
Catalog No. 503043728
Encompass_Preferred
Dynarex povidone iodine impregnated swabsticks are ideal for antiseptic skin preparation, venipuncture, IV starts, renal dialysis, pre-op prepping and other minor invasive procedures Each 4-inch-long swabstick is saturated with a 10% povidone iodine solution Antiseptic Germicide Tip is 30 millimeters wide
Catalog No. 50-304-3728 Supplier Med Vet International Supplier No. 688575BX
Dynarex povidone iodine impregnated swabsticks are ideal for antiseptic skin preparation, venipuncture, IV starts, renal dialysis, pre-op prepping and other minor invasive procedures Each 4-inch-long swabstick is saturated with a 10% povidone iodine solution Antiseptic Germicide Tip is 30 millimeters wide