Learn More
Thomas Scientific Swab-Sampler 1ml Letheen Broth, 100 case
SDP
Catalog No. NC1790881
Encompass
Swab in pre-filled bottle with various diluents. Swab reaches into corners and hard-to-reach areas for increased accuracy. Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant. Bottles have ample headspace for sample addition and vortexing. Self-contained swab and vial for increased productivity and efficiency.
Catalog No. NC1790881 Supplier Thomas Scientific Supplier No. 1185Y19
Swab in pre-filled bottle with various diluents. Swab reaches into corners and hard-to-reach areas for increased accuracy. Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant. Bottles have ample headspace for sample addition and vortexing. Self-contained swab and vial for increased productivity and efficiency.