Learn More
BIRCHWOOD LABORATORIES SWABS 16IN SCOPETTE 500/CS
Catalog No. NC1503908
Encompass
This item has a minimum qty of 2 per supplier requirements.
Uniform and highly absorbent with 5/8 inch Rayon heads and 3/16 inch cushioned tips,NON-STERILE; 16" cellulose sized swabs. 500/cs
This item is not returnable. View return policy
Catalog No. NC1503908 Supplier BIRCHWOOD LABORATORIES Supplier No. 3470238
This item has a minimum qty of 2 per supplier requirements.
Uniform and highly absorbent with 5/8 inch Rayon heads and 3/16 inch cushioned tips,NON-STERILE; 16" cellulose sized swabs. 500/cs