Learn More
Hach Company Swabs-Cotton-Presterilized
Catalog No. 5000257173 Shop All Hach Company Products
Encompass
Swabs (cotton tip hardwood stick) used to transfer colonies from positive membrane filtration plates to confirmation broth Length 6 inches
Catalog No. 50-002-57173 Supplier Hach Company Supplier No. 2554300
Swabs (cotton tip hardwood stick) used to transfer colonies from positive membrane filtration plates to confirmation broth Length 6 inches