Learn More
Med Vet International McKesson Impregnated Swabstick, 70% Isopropyl Alcohol, Sterile, 988, 750/Case
Catalog No. 503043976
Encompass_Preferred
Impregnated Swabstick McKesson 70% Strength Isopropyl Alcohol Individual Packet Sterile Swabsticks impregnated with 70% isopropyl alcohol Sterile for use on wounds and scrapes Essential first aid supply Useful for critical medical environments Each individual packet includes 3 disposable swab sticks Directions for use: Open packet and remove swab sticks; apply topically as needed; discard after single use
Catalog No. 50-304-3976 Supplier Med Vet International Supplier No. 854754CS
Impregnated Swabstick McKesson 70% Strength Isopropyl Alcohol Individual Packet Sterile Swabsticks impregnated with 70% isopropyl alcohol Sterile for use on wounds and scrapes Essential first aid supply Useful for critical medical environments Each individual packet includes 3 disposable swab sticks Directions for use: Open packet and remove swab sticks; apply topically as needed; discard after single use