Learn More
Med Vet International McKesson Nasopharyngeal Collection Swab, 5½ Inch, 1338, 500/Case
Catalog No. 503043060
Encompass_Preferred
Nasopharyngeal Collection Swab McKesson 5-1/2 Inch Length Sterile Flexible aluminum shaft Ideal specimen collection tool for testing and screening Miniature calcium alginate tip is biodegradable
Catalog No. 50-304-3060 Supplier Med Vet International Supplier No. 883891CS
Nasopharyngeal Collection Swab McKesson 5-1/2 Inch Length Sterile Flexible aluminum shaft Ideal specimen collection tool for testing and screening Miniature calcium alginate tip is biodegradable