Learn More
Med Vet International Impregnated Swabstick, 1 per Pack, Povidone-Iodine, Individual Packet, NonSterile, MDS093902, 500/Case
Catalog No. 503045465
Encompass_Preferred
Povidone Iodine kills bacteria, fungi, viruses, protozoa and yeast Forms skin protection against microbes until the solution is washed from the skin 4 inch swabsticks are non-sterile with a plastic shaft and a rayon tip
Catalog No. 50-304-5465 Supplier Med Vet International Supplier No. 370602CS
Povidone Iodine kills bacteria, fungi, viruses, protozoa and yeast Forms skin protection against microbes until the solution is washed from the skin 4 inch swabsticks are non-sterile with a plastic shaft and a rayon tip