Learn More
Med Vet International Impregnated Swabstick, 3 per Pack, Povidone-Iodine, Individual Packet, NonSterile, MDS093902, 250/Case
Catalog No. 503043693
Encompass_Preferred
Swabstick is saturated with a 10% povidone-iodine, USP solution, a broad-range, first aid antimicrobial that helps prevent infection in minor cuts, scrapes and burns Solution is equivalent to 1% available iodine This product is intended for use on the body only
Catalog No. 50-304-3693 Supplier Med Vet International Supplier No. 1085583CS
Swabstick is saturated with a 10% povidone-iodine, USP solution, a broad-range, first aid antimicrobial that helps prevent infection in minor cuts, scrapes and burns Solution is equivalent to 1% available iodine This product is intended for use on the body only