Learn More
Med Vet International McKesson Impregnated Swabstick 3 Pack 10% Strength, Sterile, 987, 750/Case
Catalog No. 503043375
Encompass_Preferred
Impregnated Swabstick McKesson 10% Strength Povidone-Iodine Individual Packet Sterile The single-use povidone swabsticks measure 4 inches in length, allowing the user to achieve a firm hold The povidone-iodine swabsticks act as a topical antiseptic and germicide They're used by doctors to prep skin prior to an injection or surgery Use them directly on minor cuts, burns and scrapes to reduce the risk of infection The sterile swabsticks come individually wrapped to prevent contamination
Catalog No. 50-304-3375 Supplier Med Vet International Supplier No. 854753CS
Impregnated Swabstick McKesson 10% Strength Povidone-Iodine Individual Packet Sterile The single-use povidone swabsticks measure 4 inches in length, allowing the user to achieve a firm hold The povidone-iodine swabsticks act as a topical antiseptic and germicide They're used by doctors to prep skin prior to an injection or surgery Use them directly on minor cuts, burns and scrapes to reduce the risk of infection The sterile swabsticks come individually wrapped to prevent contamination