Learn More
Med Vet International Dynarex Alcohol Swabsticks, 4", 500/Case
Catalog No. 502832153
Encompass_Preferred
Dynarex Alcohol Swabsticks are designed for safe and effective skin cleansing in preparation for minor medical procedures. Each antiseptic Swabstick features a soft, absorbent tip saturated with 70% isopropyl alcohol to help reduce bacteria. These premium-quality Swabsticks are packaged in convenient, easy-to-open foil packets for quick access. Ideally sized for easy portability in travel bags and first-aid kits for optimal convenience.  Dynarex Alcohol Swabsticks are convenient antiseptic swabs designed to effectively cleanse and disinfect the skin prior to procedures. These Swabsticks are saturated with 70% isopropyl alcohol to help reduce bacteria. 
Catalog No. 50-283-2153 Supplier Med Vet International Supplier No. 1203
Dynarex Alcohol Swabsticks are designed for safe and effective skin cleansing in preparation for minor medical procedures. Each antiseptic Swabstick features a soft, absorbent tip saturated with 70% isopropyl alcohol to help reduce bacteria. These premium-quality Swabsticks are packaged in convenient, easy-to-open foil packets for quick access. Ideally sized for easy portability in travel bags and first-aid kits for optimal convenience.  Dynarex Alcohol Swabsticks are convenient antiseptic swabs designed to effectively cleanse and disinfect the skin prior to procedures. These Swabsticks are saturated with 70% isopropyl alcohol to help reduce bacteria. 