Learn More
Enterprise Technology Solutions The Dynarex Povidone-Iodine Swabsticks
SDP
Catalog No. 502407230
Encompass_Preferred
The Dynarex Povidone-Iodine Swabsticks are ideal for removing dead skin, oil, and dirt from skin during skin preparation process. The swabsticks contain 10% Povidone-Iodine solution and are packaged individually or in sets of 3 swabsticks
Catalog No. 50-240-7230 Supplier Enterprise Technology Solutions Supplier No. DRX1201
The Dynarex Povidone-Iodine Swabsticks are ideal for removing dead skin, oil, and dirt from skin during skin preparation process. The swabsticks contain 10% Povidone-Iodine solution and are packaged individually or in sets of 3 swabsticks