Learn More
Med Vet International Device Swab Prevantics 3.15% / 70% Strength CHG (Chlorhexidine Gluconate) / Isopropyl Alcohol Individual Packet NonSterile, 1ML, B19600, 1000/Case
Catalog No. 503045119
Encompass_Preferred
Features a 5 second scrub time followed by 5 second dry time Contains 3.15% Chlorhexidine Gluconate and 70% Isopropyl Alcohol
Catalog No. 50-304-5119 Supplier Med Vet International Supplier No. 959981CS
Features a 5 second scrub time followed by 5 second dry time Contains 3.15% Chlorhexidine Gluconate and 70% Isopropyl Alcohol