Learn More
Med Vet International Device Swab Prevantics 3.15% / 70% Strength CHG (Chlorhexidine Gluconate) / Isopropyl Alcohol Individual Packet NonSterile, 1ML, B19600, 100/Box
Catalog No. 503045122
Encompass_Preferred
Features a 5 second scrub time followed by 5 second dry time Contains 3.15% Chlorhexidine Gluconate and 70% Isopropyl Alcohol
Catalog No. 50-304-5122 Supplier Med Vet International Supplier No. 959981BX
Features a 5 second scrub time followed by 5 second dry time Contains 3.15% Chlorhexidine Gluconate and 70% Isopropyl Alcohol