Learn More
Glass Expansion Swage Tool for Ratchet Connector
SDP
Catalog No. 502703096
Encompass_Preferred
Swage Tool for Ratchet Connector. Used to secure ferrules to capillaries at end with 1/4-28 Ratchet Connector.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3096 Supplier Glass Expansion Supplier No. 501001811
Swage Tool for Ratchet Connector. Used to secure ferrules to capillaries at end with 1/4-28 Ratchet Connector.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001