Learn More
Perkin Elmer US LLC Swagelok Brass Front Ferrule - 0.125 in Nut x 0.125 in I.D.
Catalog No. 502112695
Encompass_Preferred
Swagelok fittings are easy to install and come in a wide variety of configurations. They form gas-tight seals that are resistant to vibration fatigue and capable of withstanding high-pressures, vibrations, and temperature changes. The patented advanced-geometry back ferrule design provides a leak-tight tube connection on all Swagelok stainless steel tube fittings of identical sizes. When installed, the front ferrule creates a seal against the fitting body and on the outside diameter of the tubing. This is a replacement front ferrule for the Clarus 590/690 capillary injectors Fitting Size: 1/8 inch Ferrule Inner Diameter: 1/8 inch Corresponding Back Ferrule: (09903130)
Catalog No. 50-211-2695 Supplier Perkin Elmer US LLC Supplier No. 09903129
Swagelok fittings are easy to install and come in a wide variety of configurations. They form gas-tight seals that are resistant to vibration fatigue and capable of withstanding high-pressures, vibrations, and temperature changes. The patented advanced-geometry back ferrule design provides a leak-tight tube connection on all Swagelok stainless steel tube fittings of identical sizes. When installed, the front ferrule creates a seal against the fitting body and on the outside diameter of the tubing. This is a replacement front ferrule for the Clarus 590/690 capillary injectors Fitting Size: 1/8 inch Ferrule Inner Diameter: 1/8 inch Corresponding Back Ferrule: (09903130)