Learn More
Perkin Elmer US LLC Swagelok Brass Front Ferrules - 0.125 in Nut x 0.125 in I.D., Pkg. 5
Catalog No. 502105248
Encompass_Preferred
Swagelok fittings are easy to install and come in a wide variety of configurations. They form gas-tight seals that are resistant to vibration fatigue and capable of withstanding high-pressures, vibrations, and temperature changes. The patented advanced-geometry back ferrule design provides a leak-tight tube connection on all Swagelok stainless steel tube fittings of identical sizes. When installed, the front ferrule creates a seal against the fitting body and on the outside diameter of the tubing. Quantity: Package of 5 Fitting Size: 1/8 inch Ferrule Inner Diameter: 1/8 inch Corresponding Back Ferrules: (N9300036)
Catalog No. 50-210-5248 Supplier Perkin Elmer US LLC Supplier No. N9300037
Swagelok fittings are easy to install and come in a wide variety of configurations. They form gas-tight seals that are resistant to vibration fatigue and capable of withstanding high-pressures, vibrations, and temperature changes. The patented advanced-geometry back ferrule design provides a leak-tight tube connection on all Swagelok stainless steel tube fittings of identical sizes. When installed, the front ferrule creates a seal against the fitting body and on the outside diameter of the tubing. Quantity: Package of 5 Fitting Size: 1/8 inch Ferrule Inner Diameter: 1/8 inch Corresponding Back Ferrules: (N9300036)