Learn More
Perkin Elmer US LLC Swagelok Brass Male Adapter - 0.25 in x 0.125 in Male NPT
Catalog No. 502112391
Encompass_Preferred
Swagelok fittings are easy to install, resistant to vibration fatigue, and come in a wide variety of configurations. These leak-tight seals will withstand high-pressure, vibration, vacuum and temperature changes. Style: Male Connector Connection 1: 1/4 inch Swagelok Male Tube Adapter Connection 2: 1/8 inch Male NPT
Catalog No. 50-211-2391 Supplier Perkin Elmer US LLC Supplier No. N9301266
Swagelok fittings are easy to install, resistant to vibration fatigue, and come in a wide variety of configurations. These leak-tight seals will withstand high-pressure, vibration, vacuum and temperature changes. Style: Male Connector Connection 1: 1/4 inch Swagelok Male Tube Adapter Connection 2: 1/8 inch Male NPT