Learn More
New Jersey Fluid Systems LLC Swagelok Hose Connector, 316 Stainless Steel
Catalog No. NC9059603
Encompass
316 Stainless Steel Swagelok Hose Connector x Pipe Thread Adapter. 3/8 in. Hose Barb x 1/2 in. Male NPT
Catalog No. NC9059603 Supplier New Jersey Fluid Systems LLC Supplier No. SS6HC18
316 Stainless Steel Swagelok Hose Connector x Pipe Thread Adapter. 3/8 in. Hose Barb x 1/2 in. Male NPT