Learn More
New Jersey Fluid Systems LLC SWAGELOK MALE CONNECTOR 1/2IN
Catalog No. NC1526130
Encompass
SwagelokMale Connector, 1/2 in. Tube OD x 1/4 in. Male NPT
Catalog No. NC1526130 Supplier New Jersey Fluid Systems LLC Supplier No. SS81014
SwagelokMale Connector, 1/2 in. Tube OD x 1/4 in. Male NPT