Learn More
Perkin Elmer US LLC Swagelok PTFE Back Ferrule, 0.0625 in
Catalog No. 502117110
Encompass_Preferred
Swagelok fittings are easy to install, resistant to vibration fatigue, and come in a wide variety of configurations. These leak-tight seals will withstand high-pressure, vibration, vacuum and temperature changes. Fitting Size: 1/16 inch Ferrule Inner Diameter: 1/16 inch Corresponding Front Ferrule: (09920123)
Catalog No. 50-211-7110 Supplier Perkin Elmer US LLC Supplier No. 09920124
Swagelok fittings are easy to install, resistant to vibration fatigue, and come in a wide variety of configurations. These leak-tight seals will withstand high-pressure, vibration, vacuum and temperature changes. Fitting Size: 1/16 inch Ferrule Inner Diameter: 1/16 inch Corresponding Front Ferrule: (09920123)