Learn More
Perkin Elmer US LLC Swagelok PTFE Back Ferrule, 0.25 in
Catalog No. 502112696
Encompass_Preferred
Swagelok fittings are easy to install and come in a wide variety of configurations. They form gas-tight seals that are resistant to vibration fatigue and capable of withstanding high-pressures, vibrations, and temperature changes. The patented advanced-geometry back ferrule design provides a leak-tight tube connection on all Swagelok stainless steel tube fittings of identical sizes. When installed, the back ferrule axially advances on the front ferrule, radially applying an effective tube grip. Fitting Size: 1/4 inch Ferrule Inner Diameter: 1/4 inch Corresponding Front Ferrule: (09903119)
Catalog No. 50-211-2696 Supplier Perkin Elmer US LLC Supplier No. 09903120
Swagelok fittings are easy to install and come in a wide variety of configurations. They form gas-tight seals that are resistant to vibration fatigue and capable of withstanding high-pressures, vibrations, and temperature changes. The patented advanced-geometry back ferrule design provides a leak-tight tube connection on all Swagelok stainless steel tube fittings of identical sizes. When installed, the back ferrule axially advances on the front ferrule, radially applying an effective tube grip. Fitting Size: 1/4 inch Ferrule Inner Diameter: 1/4 inch Corresponding Front Ferrule: (09903119)