Learn More
Perkin Elmer US LLC Swagelok PTFE Front Ferrule, 0.0625 in
Catalog No. 502117113
Encompass_Preferred
Swagelok fittings are easy to install, resistant to vibration fatigue, and come in a wide variety of configurations. These leak-tight seals will withstand high-pressure, vibration, vacuum and temperature changes. Fitting Size: 1/16 inch Ferrule Inner Diameter: 1/16 inch Corresponding Back Ferrule: (09920124)
Catalog No. 50-211-7113 Supplier Perkin Elmer US LLC Supplier No. 09920123
Swagelok fittings are easy to install, resistant to vibration fatigue, and come in a wide variety of configurations. These leak-tight seals will withstand high-pressure, vibration, vacuum and temperature changes. Fitting Size: 1/16 inch Ferrule Inner Diameter: 1/16 inch Corresponding Back Ferrule: (09920124)