Learn More
New Jersey Fluid Systems LLC SWAGELOK SS-8-TA-1-4
Catalog No. NC1937849
Encompass
Stainless Steel Swagelok Tube Fitting, Male Tube Adapter, 1/2 in. Tube OD x 1/4 in. Male NPT
Catalog No. NC1937849 Supplier New Jersey Fluid Systems LLC Supplier No. SS8TA14
Stainless Steel Swagelok Tube Fitting, Male Tube Adapter, 1/2 in. Tube OD x 1/4 in. Male NPT