Learn More
New Jersey Fluid Systems LLC SWAGELOK SS-810-8-4
Catalog No. NC1937761
Encompass
Stainless Steel Swagelok Tube Fitting, Female Elbow, 1/2 in. Tube OD x 1/4 in. Female NPT
Catalog No. NC1937761 Supplier New Jersey Fluid Systems LLC Supplier No. SS81084
Stainless Steel Swagelok Tube Fitting, Female Elbow, 1/2 in. Tube OD x 1/4 in. Female NPT