Learn More
Perkin Elmer US LLC Swagelok Stainless Steel LDV Union, 0.0625 in
Catalog No. 502113734
Encompass_Preferred
Swagelok fittings are easy to install and come in a wide variety of configurations. They form gas-tight seals that are resistant to vibration fatigue and capable of withstanding high-pressures, vibrations, and temperature changes. The patented advanced-geometry back ferrule design provides a leak-tight tube connection on all Swagelok stainless steel tube fittings of identical sizes. Style: Low Dead Volume Union Connection 1: 1/16 inch Female Swagelok Tube Fitting Connection 2: 1/16 inch Female Swagelok Tube Fitting
Catalog No. 50-211-3734 Supplier Perkin Elmer US LLC Supplier No. 09903807
Swagelok fittings are easy to install and come in a wide variety of configurations. They form gas-tight seals that are resistant to vibration fatigue and capable of withstanding high-pressures, vibrations, and temperature changes. The patented advanced-geometry back ferrule design provides a leak-tight tube connection on all Swagelok stainless steel tube fittings of identical sizes. Style: Low Dead Volume Union Connection 1: 1/16 inch Female Swagelok Tube Fitting Connection 2: 1/16 inch Female Swagelok Tube Fitting