Learn More
Perkin Elmer US LLC Swagelok Stainless Steel Nut, 0.125 in
Catalog No. 502112747
Encompass_Preferred
Swagelok fittings are easy to install and come in a wide variety of configurations. They form gas-tight seals that are resistant to vibration fatigue and capable of withstanding high-pressures, vibrations, and temperature changes. The patented advanced-geometry back ferrule design provides a leak-tight tube connection on all Swagelok stainless steel tube fittings of identical sizes. Quantity: Package of 5 Style: Nut Inner Diameter: 1/4 inch
Catalog No. 50-211-2747 Supplier Perkin Elmer US LLC Supplier No. 09903453
Swagelok fittings are easy to install and come in a wide variety of configurations. They form gas-tight seals that are resistant to vibration fatigue and capable of withstanding high-pressures, vibrations, and temperature changes. The patented advanced-geometry back ferrule design provides a leak-tight tube connection on all Swagelok stainless steel tube fittings of identical sizes. Quantity: Package of 5 Style: Nut Inner Diameter: 1/4 inch