Learn More
Perkin Elmer US LLC Swagelok Stainless Steel Reducing Union - 0.25 in I.D. x 0.125 in I.D.
Catalog No. 502112378
Encompass_Preferred
Swagelok fittings are easy to install and come in a wide variety of configurations. They form gas-tight seals that are resistant to vibration fatigue and capable of withstanding high-pressures, vibrations, and temperature changes. The patented advanced-geometry back ferrule design provides a leak-tight tube connection on all Swagelok stainless steel tube fittings of identical sizes. Style: Reducing Union Connection 1: 1/4 inch Swagelok Tube Fitting Connection 2: 1/8 inch Swagelok Tube Fitting
Catalog No. 50-211-2378 Supplier Perkin Elmer US LLC Supplier No. N9301226
Swagelok fittings are easy to install and come in a wide variety of configurations. They form gas-tight seals that are resistant to vibration fatigue and capable of withstanding high-pressures, vibrations, and temperature changes. The patented advanced-geometry back ferrule design provides a leak-tight tube connection on all Swagelok stainless steel tube fittings of identical sizes. Style: Reducing Union Connection 1: 1/4 inch Swagelok Tube Fitting Connection 2: 1/8 inch Swagelok Tube Fitting