Learn More
Strem, An Ascensus Company 1cyl. Swagelok Cylinder Assembly, 50ml with 1/4" VCR Male Bellows-Sealed Valve (High Temp) and Female Nut for CVD/ALD.
SDP
Catalog No. 5090116580
Encompass
1cyl. Swagelok Cylinder Assembly, 50ml with 1/4" VCR Male Bellows-Sealed Valve (High Temp) and Female Nut for CVD/ALD. Strem# 96-1071. http://www.strem.com/catalog/v/96-1071/
Catalog No. 50-901-16580 Supplier Strem, An Ascensus Company Supplier No. 9610711CYL
1cyl. Swagelok Cylinder Assembly, 50ml with 1/4" VCR Male Bellows-Sealed Valve (High Temp) and Female Nut for CVD/ALD. Strem# 96-1071. http://www.strem.com/catalog/v/96-1071/