Learn More
New England Biolabs, Inc. SwaI – 2000 units
SDP
Catalog No. 50812330
Encompass
A restriction endonuclease that recognizes the sequence ATTT_^AAAT.
Catalog No. 50-812-330 Supplier New England Biolabs, Inc. Supplier No. R0604S
A restriction endonuclease that recognizes the sequence ATTT_^AAAT.