Learn More
Enzo Life Sciences Swainsonine (1 mg). CAS: 72741-87-8
SDP
Supplier:  Enzo Life Sciences BMLS1120001
Encompass_Preferred
Reversibly inhibits the active site of lysosomal α-mannosidase. Also inhibits the growth of tumors and prevents metastasis in murine models. Could inhibit cell proliferation in vitro and the growth of human gastric carcinoma in vivo. Blocks the processing of high mannose oligosaccharides to form complex type oligosaccharides.^Immunomodulator. Alternative name: 8alpha,beta-Octahydroindolizidine-1alpha,2alpha,8beta-triol. Purity: ≥98% (HPLC). Solubility: Soluble in 100% ethanol, DMSO, methanol or water (10mg/ml). Long Term Storage: +4°C.
Catalog No. 50-201-1737